Merck
  • Disrupted calcium homeostasis is involved in elevated zinc ion-induced photoreceptor cell death.
Disrupted calcium homeostasis is involved in elevated zinc ion-induced photoreceptor cell death.
Archives of biochemistry and biophysics (2014-07-23)
Dadong Guo, Yuxiang Du, Qiuxin Wu, Wenjun Jiang, Hongsheng Bi
ABSTRACT
Zinc (Zn), the second abundant trace element in living organisms, plays an important role in regulating cell metabolism, signaling, proliferation, gene expression and apoptosis. Meanwhile, the overload of Zn will disrupt the intracellular calcium homeostasis via impairing mitochondrial function. However, the specific molecular mechanism underlying zinc-induced calcium regulation remains poorly understood. In the present study, using zinc chloride (ZnCl2) as a stressor, we investigated the effect of exogenous Zn(2+) in regulating murine photoreceptor cell viability, reactive oxygen species (ROS), cell cycle distribution and calcium homeostasis as well as plasma membrane calcium ATPase (PMCA) isoforms (PMCA1 and PMCA2, i.e., ATP2B1, ATP2B2) expression. We found that the exogenous Zn(2+) in the exposure range (31.25-125.0 μmol/L) results in the overgeneration of ROS, cell cycle arrest at G2/M phases, elevation of cytosolic [Ca(2+)], inactivation of Ca(2+)-ATPase and reduction of both PMCA1 and PMCA2 in 661 W cells, and thus induces cell death. In conclusion, ZnCl2 exposure can elevate the cytosolic [Ca(2+)], disrupt the intracellular calcium homeostasis, further initiate Ca(2+)-dependent signaling pathway in 661 W cells, and finally cause cell death. Our results will facilitate the understanding of cell death induced by the zinc ion-mediated calcium homeostasis disruption.
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
Propidium iodide solution