Merck
  • Medication-associated diethylene glycol mass poisoning: a review and discussion on the origin of contamination.
Medication-associated diethylene glycol mass poisoning: a review and discussion on the origin of contamination.
Journal of public health policy (2009-07-15)
Joshua G Schier, Carol S Rubin, Dorothy Miller, Dana Barr, Michael A McGeehin
ABSTRACT
Diethylene glycol (DEG), an extremely toxic chemical, has been implicated as the etiologic agent in at least 12 medication-associated mass poisonings over the last 70 years. Why DEG mass poisonings occur remains unclear. Most reports do not contain detailed reports of trace-back investigations into the etiology. The authors, therefore, conducted a systematic literature review on potential etiologies of these mass poisonings. The current available evidence suggests that substitution of DEG or DEG-containing compounds for pharmaceutical ingredients results from: (1) deception as to the true nature of certain ingredients by persons at some point in the pharmaceutical manufacturing process, and (2) failure to adhere to standardized quality control procedures in manufacturing pharmaceutical products intended for consumers. We discuss existing guidelines and new recommendations for prevention of these incidents.
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Supelco
Diethylene glycol, analytical standard
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
USP
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard