Merck
  • Systematic review: impact of non-adherence to 5-aminosalicylic acid products on the frequency and cost of ulcerative colitis flares.
Systematic review: impact of non-adherence to 5-aminosalicylic acid products on the frequency and cost of ulcerative colitis flares.
Alimentary pharmacology & therapeutics (2008-10-24)
P D R Higgins, D T Rubin, K Kaulback, P S Schoenfield, S V Kane
ABSTRACT
Ulcerative colitis (UC) can be maintained in remission with 5-aminosalicylic acid (5-ASA) medications, but frequent non-adherence by patients who are feeling well has been associated with more frequent flares of colitis. To perform a systematic review of the published literature and unpublished randomized clinical trials (RCTs) regarding the impact of non-adherence with 5-ASA medications on the incidence of UC flares and costs of care. A search of MEDLINE, EMBASE and the Cochrane databases was performed. Prospective studies of UC maintenance with 5-ASAs in adults were selected if they included data on adherence and disease flares. Studies using insurance claims data to estimate the impact of non-adherence on cost of care were included. Data from unpublished RCTs were obtained from the FDA with a request under the Freedom of Information Act. The relative risk for flare in non-adherent vs. adherent patients ranged from 3.65 to infinity. Data were obtained from six unpublished 5-ASA RCTs, but none measured the impact of adherence on disease activity. The comorbidity-adjusted annual costs of care in adherent patients were 12.5% less than in non-adherent patients, despite increased medication expenditures. A substantial proportion of UC flares and medical costs of UC are attributable to 5-ASA non-adherence. As non-adherence to 5-ASA medications is common, cost-effective strategies to improve adherence are needed. The impact of adherence on disease activity should be measured in RCTs of all inflammatory bowel disease treatments.
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
Mesalazine, European Pharmacopoeia (EP) Reference Standard
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard