Merck
  • Pharmacodynamic interaction of naproxen with low-dose aspirin in healthy subjects.
Pharmacodynamic interaction of naproxen with low-dose aspirin in healthy subjects.
Journal of the American College of Cardiology (2005-04-20)
Marta L Capone, Maria G Sciulli, Stefania Tacconelli, Marilena Grana, Emanuela Ricciotti, Giulia Renda, Patrizia Di Gregorio, Gabriele Merciaro, Paola Patrignani
ABSTRACT
We investigated the occurrence of pharmacodynamic interaction between low-dose aspirin and naproxen. The uncertainty of cardioprotection by naproxen has encouraged its combination with aspirin in patients with arthritis and cardiovascular disease. The incubation of washed platelets with naproxen for 5 min before the addition of aspirin reduced the irreversible inhibition of thromboxane (TX)B(2) production by aspirin. The pharmacodynamic interaction between the two drugs was then investigated in four healthy volunteers who received aspirin (100 mg daily) for 6 days and then the combination of aspirin and naproxen for further 6 days: aspirin 2 h before naproxen (500 mg, twice-daily dosing). After 14 days of washout, naproxen was given 2 h before aspirin for further 6 days. The inhibition of serum TXB(2) production (index of platelet cyclooxygenase [COX]-1 activity) and platelet aggregation ex vivo and urinary 11-dehydro-TXB(2) levels (index of TXB(2) biosynthesis in vivo) by aspirin alone (99 +/- 0.2%, 95 +/- 0.6%, and 81 +/- 4%, respectively) was not significantly altered by the co-administration of naproxen, given either 2 h after aspirin or in reverse order. In a second study, the concurrent administration of a single dose of aspirin and naproxen did not affect platelet TXB(2) production and aggregation at 1 h after dosing, when aspirin alone causes maximal inhibitory effect. Moreover, the rapid recovery of platelet COX-1 activity and function supports the occurrence of a pharmacodynamic interaction between naproxen and aspirin. Naproxen interfered with the inhibitory effect of aspirin on platelet COX-1 activity and function. This pharmacodynamic interaction might undermine the sustained inhibition of platelet COX-1 that is necessary for aspirin's cardioprotective effects.
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
Naproxen, VETRANAL®, analytical standard
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Naproxen, European Pharmacopoeia (EP) Reference Standard
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard
Supelco
Naproxen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®