Merck
What do we know about taper corrosion in total hip arthroplasty?
The Journal of arthroplasty (2014-03-25)
J J Jacobs, H J Cooper, R M Urban, R L Wixson, C J Della Valle
ABSTRACT
Mechanically assisted crevice corrosion (MACC) at metal/metal modular junctions in which at least one of the components is fabricated from cobalt-chromium alloy, has reemerged as a potential clinically significant complication in total hip arthroplasty. The clinical manifestation of MACC may include the development of an adverse local tissue reaction (ALTR), similar to what has been described in association with metal-on-metal bearing total hip and resurfacing arthroplasty. The clinical presentation of MACC-associated ALTRs may include pain and possibly late recurrent dislocations. Abnormal metal artifact reduction sequence magnetic resonance images and elevated serum metal levels (cobalt elevations out of proportion to chromium elevations) can be helpful in the diagnosis of these MACC-associated ALTRs.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Cobalt, foil, 6mm disks, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%
Cobalt, wire reel, 0.05m, diameter 1.0mm, hard, 99.99+%
Cobalt, foil, light tested, 25x25mm, thickness 0.025mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.0025mm, temporary acrylic support, 99.9%
Cobalt, foil, light tested, 50x50mm, thickness 0.015mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 8mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.02mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.9%
Cobalt, wire reel, 0.05m, diameter 2.0mm, hard, 99.99+%
Cobalt, foil, not light tested, 25x25mm, thickness 0.002mm, temporary acrylic support, 99.9%