Merck
  • Multicolor photoswitching microscopy for subdiffraction-resolution fluorescence imaging.
Multicolor photoswitching microscopy for subdiffraction-resolution fluorescence imaging.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2009-04-02)
Sebastian van de Linde, Ulrike Endesfelder, Anindita Mukherjee, Mark Schüttpelz, Gerd Wiebusch, Steve Wolter, Mike Heilemann, Markus Sauer
ABSTRACT
We introduce a general approach for multicolor subdiffraction-resolution fluorescence imaging based on photoswitching of standard organic fluorophores. Photoswitching of ordinary fluorophores such as ATTO520, ATTO565, ATTO655, ATTO680, or ATTO700, i.e. the reversible transition from a fluorescent to a nonfluorescent state in aqueous buffers exploits the formation of long-lived triplet radical anions through reaction with reducing agents such as beta-mercaptoethylamine and repopulation of the singlet ground state by interaction with molecular oxygen. Thus, the time the different fluorophores reside in the fluorescent state can be easily adjusted by the excitation intensity and the concentration of the reducing agent. We demonstrate the potential of multicolor photoswitching microscopy with subdiffraction-resolution on cytoskeletal networks and molecular quantification of proteins in the inner mitochondrial membrane with approximately 20 nm optical resolution.
Sigma-Aldrich
Atto 565, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 655-Biotin, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 565-Biotin
Sigma-Aldrich
Atto 655, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Atto 655-maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling rate)
Sigma-Aldrich
Atto 565 maleimide, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 655 NHS ester, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 700 NHS-ester, BioReagent, suitable for fluorescence, ≥90% (coupling to amine, HPLC)
Sigma-Aldrich
Atto 700, BioReagent, suitable for fluorescence, ≥90.0% (HPCE)
Supelco
Atto 565 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)