Merck
  • Serotonin and aversive Pavlovian control of instrumental behavior in humans.
Serotonin and aversive Pavlovian control of instrumental behavior in humans.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-11-29)
Dirk E M Geurts, Quentin J M Huys, Hanneke E M den Ouden, Roshan Cools
ABSTRACT
Adaptive decision-making involves interaction between systems regulating Pavlovian and instrumental control of behavior. Here we investigate in humans the role of serotonin in such Pavlovian-instrumental transfer in both the aversive and the appetitive domain using acute tryptophan depletion, known to lower central serotonin levels. Acute tryptophan depletion attenuated the inhibiting effect of aversive Pavlovian cues on instrumental behavior, while leaving unaltered the activating effect of appetitive Pavlovian cues. These data suggest that serotonin is selectively involved in Pavlovian inhibition due to aversive expectations and have implications for our understanding of the mechanisms underlying a range of affective, impulsive, and aggressive neuropsychiatric disorders.
Product Description
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)