Merck
  • Development of mucoadhesive patches for buccal administration of ibuprofen.
Development of mucoadhesive patches for buccal administration of ibuprofen.
Journal of controlled release : official journal of the Controlled Release Society (2004-09-03)
Luana Perioli, Valeria Ambrogi, Fausta Angelici, Maurizio Ricci, Stefano Giovagnoli, Marinella Capuccella, Carlo Rossi
ABSTRACT
A new formulation for topical administration of drugs in the oral cavity has been developed using several film-forming and mucoadhesive polymers. The films have been evaluated in terms of swelling, mucoadhesion and organoleptic characteristics. The best film, containing polyvinylpyrrolidone (PVP) as film-forming polymer and carboxymethylcellulose sodium salt (NaCMC) as mucoadhesive polymer, was loaded with ibuprofen as a model compound and in vitro and in vivo release studies were performed. Statistical investigation of in vitro release revealed that the diffusion process was the main drug release mechanism and the Higuchi's model provided the best fit. In vivo studies showed the presence of ibuprofen in saliva (range 70-210 microg/ml) for 5 h and no irritation was observed. These mucoadhesive formulations offer many advantages in comparison to traditional treatments and can be proposed as a new therapeutic tool against dental and buccal diseases and disturbs.
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®