Merck
  • Potential therapy of multidrug-resistant and extremely drug-resistant tuberculosis with thioridazine.
Potential therapy of multidrug-resistant and extremely drug-resistant tuberculosis with thioridazine.
In vivo (Athens, Greece) (2012-02-22)
Leonard Amaral, Joseph Molnar
ABSTRACT
Multidrug-resistant tuberculosis (MDRTB) infections that continue to increase in frequency globally have progressed to become extremely drug-resistant tuberculosis (XDRTB). The therapeutic problems associated with MDRTB pale in comparison to those for XDRTB where mortality is high. This mini-review highlights the evidence that supports the use of the phenothiazine neuroleptic thioridazine for the therapy of XDRTB. Although thioridazine does produce some serious side-effects, the poor prognosis associated with an XDRTB infection of a patient that presents with AIDS merits that the use of thioridazine for therapy of XDRTB is seriously considered. A recommended protocol is presented.
Product Description
Supelco
Thioridazine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Thioridazine for system suitability, European Pharmacopoeia (EP) Reference Standard