Merck
  • Fluoride reversibly blocks HVA calcium current in mammalian thalamic neurones.
Fluoride reversibly blocks HVA calcium current in mammalian thalamic neurones.
Neuroreport (1994-01-31)
L Bertollini, G Biella, E Wanke, G Avanzini, M de Curtis
ABSTRACT
The effect of intracellular fluoride ions on voltage-dependent calcium currents was tested during whole-cell voltage-clamp recordings in thalamic neurones acutely dissociated from young adult rats. It is demonstrated that 5-30 mM intracellular fluoride selectively and reversibly suppresses the high voltage-activated, dihydropyridine-sensitive calcium current, without affecting the transient, low voltage-activated calcium current. Intracellular diffusion of a fluoride-free solution restores the blocking effect on the slow inactivating current induced by a transitory fluoride perfusion obtained by filling the patch microelectrode tip with caesium fluoride.
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis