Merck
  • A modified, high yield procedure for the synthesis of unlabeled and 14C-labeled 4-methylene-DL-glutamic acid.
A modified, high yield procedure for the synthesis of unlabeled and 14C-labeled 4-methylene-DL-glutamic acid.
Preparative biochemistry (1981-01-01)
G K Powell, E E Dekker
ABSTRACT
This paper describes the complete chemical synthesis of 4-methylene-DL-glutamic acid from diethylmalonate, formaldehyde and diethyl acetamidomalonate. The amino acid was obtained pure following ion-exchange chromatography and/or crystallization from hot water in an overall yield of 30% based on the amount of diethylmalonate used. Several physico-chemical characteristics of the synthetic compound were determined, including ir and pmr spectra, chromatography on paper, retention time on an amino acid analyzer, pK values and melting point; all properties of the synthetic material were found to be identical to those seen with the naturally occurring L-isomer. The procedure for obtaining gram quantities of the unlabeled compound has also been modified for the synthesis of high specific activity (10.6 mCi/mol) 4-methylene-[2-14C]-DL-glutamic acid.
Sigma-Aldrich
Diethylmalonic acid, 98%
Sigma-Aldrich
Diethyl acetamidomalonate-2-13C, ≥99 atom % 13C, ≥99% (CP)
Sigma-Aldrich
Diethyl acetamidomalonate, 98%