Merck
Approaching two-dimensional polymers with macroscopic sizes.
Chimia (2013-08-24)
Payam Payamyar, Junji Sakamoto, A Dieter Schlüter
ABSTRACT
We describe the challenges involved with extending the limited lateral size of two-dimensional polymers (2DPs). An amphiphilic monomer with three-fold symmetry is chosen to form an ideally tessellated monolayer at the air/water interface. Anthracene [4+4] photo-dimerization is chosen as the growth reaction. Formation of covalent net-points upon anthracene dimerization has an influence on the mechanical coherence of the resulting sheets which could be investigated qualitatively and quantitatively by means of AFM nano-indentation.
Product Description
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, reagent grade, 97%
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland