Merck
  • Self-assembly of an asymmetrically functionalized [6]helicene at liquid/solid interfaces.
Self-assembly of an asymmetrically functionalized [6]helicene at liquid/solid interfaces.
Chemical communications (Cambridge, England) (2013-02-12)
Tatyana Balandina, Maarten W van der Meijden, Oleksandr Ivasenko, David Cornil, Jérôme Cornil, Roberto Lazzaroni, Richard M Kellogg, Steven De Feyter
ABSTRACT
STM brings to light chirality aspects of the self-assembly of a functionalized helicene at the interface between a liquid and the solid substrates, gold and graphite. This reveals conditions for conglomerate formation.
Sigma-Aldrich
1,2,4-Trichlorobenzene, anhydrous, ≥99%
Supelco
1,2,4-Trichlorobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
1,2,4-Trichlorobenzene, suitable for HPLC, ≥99%
Sigma-Aldrich
1,2,4-Trichlorobenzene, ReagentPlus®, ≥99%