Merck
  • Production and characterization of a bioflocculant produced by Aspergillus flavus.
Production and characterization of a bioflocculant produced by Aspergillus flavus.
Bioresource technology (2012-11-20)
Ahmad H Rajab Aljuboori, Azni Idris, Norhafizah Abdullah, Rosfarizan Mohamad
ABSTRACT
The production and characterization of a bioflocculant, IH-7, by Aspergillus flavus was investigated. About 0.4 g of purified bioflocculant with an average molecular weight of 2.574 × 10(4)Da could be obtained from 1L of fermentation medium. The bioflocculant mainly consisted of protein (28.5%) and sugar (69.7%), including 40% of neutral sugar, 2.48% of uronic acid and 1.8% amino sugar. The neutral sugar components are sucrose, lactose, glucose, xylose, galactose, mannose and fructose at a molar ratio of 2.4:4.4:4.1:5.8:9.9:0.8:3.1. Fourier-transform infrared spectroscopy analysis revealed that purified IH-7 contained hydroxyl, amide, carboxyl and methoxyl groups. The elemental analysis of purified IH-7 showed that the weight fractions of the elements C, H, O, N and S were 29.9%, 4.8%, 34.7%, 3.3%, and 2.0%, respectively. IH-7 had good flocculating rate in kaolin suspension without cation addition and stable over wide range of pH and temperature.
Sigma-Aldrich
Halloysite nanoclay
Sigma-Aldrich
Kaolin, tested according to Ph. Eur.
Sigma-Aldrich
Kaolin, heavy, powder
Sigma-Aldrich
Kaolin, meets USP testing specifications
Sigma-Aldrich
Kaolin