Merck
  • Electrochemical oxidation of electrodialysed reverse osmosis concentrate on Ti/Pt-IrO2, Ti/SnO2-Sb and boron-doped diamond electrodes.
Electrochemical oxidation of electrodialysed reverse osmosis concentrate on Ti/Pt-IrO2, Ti/SnO2-Sb and boron-doped diamond electrodes.
Water research (2012-11-10)
Arseto Y Bagastyo, Damien J Batstone, Korneel Rabaey, Jelena Radjenovic
ABSTRACT
Reverse osmosis concentrate from wastewater reclamation contains biorefractory trace organic contaminants that may pose environmental or health hazard. Due to its high conductivity, electrochemical oxidation of brine requires low voltage which is energetically favourable. However, the presence of chloride ions may lead to the formation of chlorinated by-products, which are likely to exert an increased toxicity and persistence to further oxidation than their non-chlorinated analogues. Here, the performance of Ti/Pt-IrO(2), Ti/SnO(2)-Sb and Si/BDD anodes was evaluated for the electrochemical oxidation of ROC in the presence of chloride, nitrate or sulfate ions (0.05 M sodium salts). In order to investigate the electrooxidation of ROC with nitrate and sulfate ions as dominant ion mediators, chloride ion concentration was decreased 10 times by electrodialytic pretreatment. The highest Coulombic efficiency for chemical oxygen demand (COD) removal was observed in the presence of high chloride ions concentration for all anodes tested (8.3-15.9%). Electrooxidation of the electrodialysed concentrate at Ti/SnO(2)-Sb and Ti/Pt-IrO(2) electrodes exhibited low dissolved organic carbon (DOC) (i.e. 23 and 12%, respectively) and COD removal (i.e. 37-43 and 6-22%, respectively), indicating that for these electrodes chlorine-mediated oxidation was the main oxidation mechanism, particularly in the latter case. In contrast, DOC removal for the electrodialysed concentrate stream was enhanced at Si/BDD anode in the presence of SO(4)(2-) (i.e. 51%) compared to NO(3)(2-) electrolyte (i.e. 41%), likely due to the contribution of SO(4)(·-) and S(2)O(8)(2-) species to the oxidative degradation. Furthermore, decreased concentration of chloride ions lead to a lower formation of haloacetic acids and trihalomethanes at all three electrodes tested.
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium nitrate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium bisulfate monohydrate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium sulfate decahydrate, BioUltra, ≥99.0% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bisulfate monohydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate decahydrate, puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Sigma-Aldrich
Sodium sulfate decahydrate, reagent grade, 97%
Sigma-Aldrich
Sodium sulfate decahydrate, ACS reagent, ≥99.0%