Merck
Mechanical activation of CaO-based adsorbents for CO(2) capture.
ChemSusChem (2012-11-08)
Maryam Sayyah, Yongqi Lu, Richard I Masel, Kenneth S Suslick
ABSTRACT
The reversible cycling of CaO adsorbents to CaCO(3) for high-temperature CO(2) capture is substantially improved by mechanical treatment. The mechanical milling intensity and conditions of grinding (e.g., wet vs. dry, planetary vs. vibratory milling) were determined to be the main factors that control the effectiveness of the mechanochemical synthesis to enhance the recycling stability of the sorbents prepared. In addition, MgO was used as an example of an inert binder to help mitigate CaCO(3) sintering. Wet planetary milling of MgO into CaCO(3) allowed efficient particle size reduction and the effective dispersion of MgO throughout the particles. Wet planetary milling yielded the most stable sorbents during 50 cycles of carbonation-calcination.
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Calcium oxide, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium oxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium oxide, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Calcium oxide, nanopowder, <160 nm particle size (BET), 98%
Sigma-Aldrich
Calcium oxide, reagent grade