Merck
  • Triple-helical recognition of RNA using 2-aminopyridine-modified PNA at physiologically relevant conditions.
Triple-helical recognition of RNA using 2-aminopyridine-modified PNA at physiologically relevant conditions.
Angewandte Chemie (International ed. in English) (2012-11-06)
Thomas Zengeya, Pankaj Gupta, Eriks Rozners
ABSTRACT
Peptide nucleic acids containing thymidine and 2-aminopyridine (M) nucleobases form stable and sequence-selective triple helices with double-stranded RNA at physiologically relevant conditions. The M-modified PNA showed unique RNA selectivity by having two orders of magnitude higher affinity for the double-stranded RNAs than for the same DNA sequences.
Sigma-Aldrich
2-Aminopyridine, ≥99%
Sigma-Aldrich
2-Aminopyridine, purum, ≥98.0% (NT)
Supelco
2-Aminopyridine, PESTANAL®, analytical standard
Sigma-Aldrich
2-Aminopyridine, 99%