Merck
  • Design, synthesis and evaluation of N-acetyl glucosamine (NAG)-PEG-doxorubicin targeted conjugates for anticancer delivery.
Design, synthesis and evaluation of N-acetyl glucosamine (NAG)-PEG-doxorubicin targeted conjugates for anticancer delivery.
International journal of pharmaceutics (2012-06-23)
Smita K Pawar, Archana J Badhwar, Firuza Kharas, Jayant J Khandare, Pradeep R Vavia
ABSTRACT
Efficacy of anticancer drug is limited by the severe adverse effects induced by drug; therefore the crux is in designing delivery systems targeted only to cancer cells. Toward this objectives, we propose, synthesis of poly(ethylene glycol) (PEG)-doxorubicin (DOX) prodrug conjugates consisting N-acetyl glucosamine (NAG) as a targeting moiety. Multicomponent system proposed here is characterized by (1)H NMR, UV spectroscopy, and HPLC. The multicomponent system is evaluated for in vitro cellular kinetics and anticancer activity using MCF-7 and MDA-MB-231 cells. Molecular modeling study demonstrated sterically stabilized conformations of polymeric conjugates. Interestingly, PEG-DOX conjugate with NAG ligand showed significantly higher cytotoxicity compared to drug conjugate with DOX. In addition, the polymer drug conjugate with NAG and DOX showed enhanced internalization and retention effect in cancer cells, compared to free DOX. Thus, with enhanced internalization and targeting ability of PEG conjugate of NAG-DOX has implication in targeted anticancer therapy.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350