Merck
  • [Volatile metabolites analysis and molecular identification of endophytic fungi bn12 from Cinnamomum camphora chvar. borneol].
[Volatile metabolites analysis and molecular identification of endophytic fungi bn12 from Cinnamomum camphora chvar. borneol].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-03-08)
Meilan Chen, Li Yang, Qin Li, Ye Shen, Aijuan Shao, Shufang Lin, Luqi Huang
ABSTRACT
To identify endophytic fungi bn12 from Cinnamomum camphora chvar, borneol and analysis its volatile metabolites. The endophytic fungi bn12 was identified by morphological observation. volatile metabolites of endophytic fungi bn12 was analyzed by gas chromatography/mass spectrography (GC-MS). Volatile metabolites of endophytic fungi bn12 contain borneol and much indoles. The ITS sequence of endophytic fungi bnl2 is most similar to the ITS sequence of pleosporaceae fungus, particularly C. nisikadoi. Endophytic fungi bn12 is belong to pleosporaceae fungus. It has the ability of producing broneol.
Sigma-Aldrich
Isoborneol, 95%
Sigma-Aldrich
(−)-Borneol, predominantly endo, 97%
Supelco
(−)-Borneol, analytical standard
Supelco
(+)-Borneol, analytical standard
Sigma-Aldrich
(+)-Borneol, 97%