Merck
  • Synthesis and luminescence properties of lanthanide complexes with a new tripodal ligands featuring salicylamide arms.
Synthesis and luminescence properties of lanthanide complexes with a new tripodal ligands featuring salicylamide arms.
Luminescence : the journal of biological and chemical luminescence (2009-09-11)
Xue-Qin Song, Wen-Kui Dong, Yu-Jie Zhang, Wei-Sheng Liu
ABSTRACT
A series of luminescent lanthanide complexes with a new tripodal ligand featuring salicylamide arms, 2,2',2''-nitrilotris(2-furfurylaminoformylphenoxy)triethylamine (L), were synthesized and characterized by elemental analysis, IR and molar conductivity measurements. Photophysical properties of the complexes were studied by means of UV-vis absorption and steady-state luminescence spectroscopy. Excited-state luminescence lifetimes and quantum yield of the complexes were determined. Luminescence studies demonstrated that the tripodal ligand featuring salicylamide arms exhibits a good antennae effect with respect to the Tb(III) and Dy(III) ion due to efficient intersystem crossing and ligand to metal energy transfer. From a more general perspective, this work offers interesting perspectives for the development of efficient luminescent stains and enlarges the arsenal for developing novel luminescent lanthanide complexes of salicylamide derivatives.
Sigma-Aldrich
Salicylamide, 99%
Sigma-Aldrich
Salicylamide, puriss., ≥99.0% (T)