Merck
  • A general approach to silicate nanoshells: gadolinium silicate and gadolinium silicate:europium nanoshells for dual-modality optical and MR imaging.
A general approach to silicate nanoshells: gadolinium silicate and gadolinium silicate:europium nanoshells for dual-modality optical and MR imaging.
Chemical communications (Cambridge, England) (2009-06-09)
Chih-Chia Huang, Wileen Huang, Chia-Hao Su, Chieh-Ning Feng, Wen-Shuo Kuo, Chen-Sheng Yeh
ABSTRACT
A general approach involving a solvothermal method was developed to synthesize a series of silicate nanoshells (<100 nm) where both Gd silicate and Gd silicate:Eu nanoshells were further demonstrated to exhibit dual-modality MRI and optical imaging functions.
Sigma-Aldrich
Manganese(II) acetylacetonate
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
Sigma-Aldrich
Copper(II) acetylacetonate, 97%
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis