Merck
  • Determinants and Mechanisms of the Low Fusogenicity and High Dependence on Endosomal Entry of Omicron Subvariants.
Determinants and Mechanisms of the Low Fusogenicity and High Dependence on Endosomal Entry of Omicron Subvariants.
mBio (2023-01-11)
Panke Qu, John P Evans, Chaitanya Kurhade, Cong Zeng, Yi-Min Zheng, Kai Xu, Pei-Yong Shi, Xuping Xie, Shan-Lu Liu
ABSTRACT
The rapid spread and strong immune evasion of the SARS-CoV-2 Omicron subvariants has raised serious concerns for the global COVID-19 pandemic. These new variants exhibit generally reduced fusogenicity and increased endosomal entry pathway utilization compared to the ancestral D614G variant, the underlying mechanisms of which remain elusive. Here, we show that the C-terminal S1 mutations of the BA.1.1 subvariant, H655Y and T547K, critically govern the low fusogenicity of Omicron. Notably, H655Y also dictates the enhanced endosome entry pathway utilization. Mechanistically, T547K and H655Y likely stabilize the spike trimer conformation as suggested by increased molecular interactions in structural modeling and enhanced S1 shedding of their reversion mutants K547T and Y655H in viral producer cells. Importantly, the H655Y mutation also determines the low fusogenicity and enhanced dependence on the endosomal entry pathway of other Omicron subvariants, including BA.2, BA.2.12.1, BA.4/5, and BA.2.75. Together, these results uncover mechanisms governing Omicron subvariant entry and provide insights into altered Omicron tissue tropism and pathogenesis. IMPORTANCE Omicron has been shown to predominantly use the endosomal entry pathway, resulting in reduced lung tropism and reduced disease severity; however, the underlying mechanism is not fully understood. In addition, whether the most recent Omicron subvariants, including BA.5 and BA.2.75, use the same pathway as their ancestor for entry is currently not known. In this study, we show that T547K and H655Y mutations in the C terminus of the S1 subunit critically determine the enhanced dependence on the endosomal entry pathway as well as the reduced cell-cell fusion activity of Omicron BA.1, BA.1.1, and other subvariants. Further experiments and molecular modeling suggest that H655Y and K547T stabilize the spike trimer conformation, likely contributing to the decreased fusogenicity and endosomal entry. Our work uncovers novel mechanisms underlying the distinct entry pathway of Omicron subvariants and advances our understanding of their biological characteristics.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
EST, A cell-permeable, irreversible inhibitor of cysteine proteases.
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Camostat mesylate, ≥98% (HPLC)