Merck
  • DDX47, MeCP2, and other functionally heterogeneous factors protect cells from harmful R loops.
DDX47, MeCP2, and other functionally heterogeneous factors protect cells from harmful R loops.
Cell reports (2023-02-25)
Esther Marchena-Cruz, Lola P Camino, Jay Bhandari, Sónia Silva, José Javier Marqueta-Gracia, Shahad A Amdeen, Cristina Guillén-Mendoza, María L García-Rubio, José M Calderón-Montaño, Xiaoyu Xue, Rosa Luna, Andrés Aguilera
ABSTRACT
Unscheduled R loops can be a source of genome instability, a hallmark of cancer cells. Although targeted proteomic approaches and cellular analysis of specific mutants have uncovered factors potentially involved in R-loop homeostasis, we report a more open screening of factors whose depletion causes R loops based on the ability of activation-induced cytidine deaminase (AID) to target R loops. Immunofluorescence analysis of γH2AX caused by small interfering RNAs (siRNAs) covering 3,205 protein-coding genes identifies 59 potential candidates, from which 13 are analyzed further and show a significant increase of R loops. Such candidates are enriched in factors involved in chromatin, transcription, and RNA biogenesis and other processes. A more focused study shows that the DDX47 helicase is an R-loop resolvase, whereas the MeCP2 methyl-CpG-binding protein uncovers a link between DNA methylation and R loops. Thus, our results suggest that a plethora of gene dysfunctions can alter cell physiology via affecting R-loop homeostasis by different mechanisms.
Sigma-Aldrich
Anti-DDX47 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
α-Amanitin, from Amanita phalloides, ≥85% (HPLC), powder
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
IgG from mouse serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture