Merck
  • ADAR1-mediated RNA editing of SCD1 drives drug resistance and self-renewal in gastric cancer.
ADAR1-mediated RNA editing of SCD1 drives drug resistance and self-renewal in gastric cancer.
Nature communications (2023-05-20)
Tin-Lok Wong, Jia-Jian Loh, Shixun Lu, Helen H N Yan, Hoi Cheong Siu, Ren Xi, Dessy Chan, Max J F Kam, Lei Zhou, Man Tong, John A Copland, Leilei Chen, Jing-Ping Yun, Suet Yi Leung, Stephanie Ma
ABSTRACT
Targetable drivers governing 5-fluorouracil and cisplatin (5FU + CDDP) resistance remain elusive due to the paucity of physiologically and therapeutically relevant models. Here, we establish 5FU + CDDP resistant intestinal subtype GC patient-derived organoid lines. JAK/STAT signaling and its downstream, adenosine deaminases acting on RNA 1 (ADAR1), are shown to be concomitantly upregulated in the resistant lines. ADAR1 confers chemoresistance and self-renewal in an RNA editing-dependent manner. WES coupled with RNA-seq identify enrichment of hyper-edited lipid metabolism genes in the resistant lines. Mechanistically, ADAR1-mediated A-to-I editing on 3'UTR of stearoyl-CoA desaturase (SCD1) increases binding of KH domain-containing, RNA-binding, signal transduction-associated 1 (KHDRBS1), thereby augmenting SCD1 mRNA stability. Consequently, SCD1 facilitates lipid droplet formation to alleviate chemotherapy-induced ER stress and enhances self-renewal through increasing β-catenin expression. Pharmacological inhibition of SCD1 abrogates chemoresistance and tumor-initiating cell frequency. Clinically, high proteomic level of ADAR1 and SCD1, or high SCD1 editing/ADAR1 mRNA signature score predicts a worse prognosis. Together, we unveil a potential target to circumvent chemoresistance.
Sigma-Aldrich
Anti-ADARB1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
A922500