Merck
  • Indirect photometric detection of polyamines in biological samples separated by high-performance capillary electrophoresis.
Indirect photometric detection of polyamines in biological samples separated by high-performance capillary electrophoresis.
Journal of chromatography (1992-09-11)
Y Ma, R Zhang, C L Cooper
ABSTRACT
A rapid separation of polyamines and some related amino acids in cultured tumor cells by high-performance capillary zone electrophoresis with indirect photometric detection is demonstrated. 60 cm x 75 microns I.D. fused-silica capillary was used for the separation and quinine sulfate was used as a background electrolyte (BGE). Several polyamines (putrescine, spermidine and spermine), amino acids (lysine, arginine, histidine) and simple cations (K+, Na+) were easily separated in less than 10 min. Using the indirect photometric detection method, femtomole amounts of polyamines extracted from the tumor cells were detected from nanoliter injection volumes, and the signal response was linear over two orders of magnitude.
Sigma-Aldrich
Quinine hemisulfate salt monohydrate, BioReagent, suitable for fluorescence, 99.0-101.0%
Pricing and availability is not currently available.