Merck
  • Reactive Oxygen Species Contributes to Type 2 Diabetic Neuropathic Pain via the Thioredoxin-Interacting Protein-NOD-Like Receptor Protein 3- N -Methyl-D-Aspartic Acid Receptor 2B Pathway.
Reactive Oxygen Species Contributes to Type 2 Diabetic Neuropathic Pain via the Thioredoxin-Interacting Protein-NOD-Like Receptor Protein 3- N -Methyl-D-Aspartic Acid Receptor 2B Pathway.
Anesthesia and analgesia (2022-07-13)
Jun-Wu Wang, Xiu-Ying Ye, Ning Wei, Shi-Shu Wu, Zhe-Hao Zhang, Guang-Hui Luo, Xu Li, Jun Li, Hong Cao
ABSTRACT
The number of patients with diabetic neuropathic pain (DNP) continues to increase, but available treatments are limited. This study aimed to examine the influence of reactive oxygen species (ROS)-thioredoxin-interacting protein (TXNIP)-NOD-like receptor protein 3 (NLRP3)- N -methyl-D-aspartic acid receptor 2B (NR2B) pathway on type 2 DNP. Male Sprague-Dawley rats were fed with a high-fat and high-sugar diet for 8 weeks. Then, rats were intraperitoneally injected with streptozotocin (STZ, 35 mg/kg) to induce type 2 diabetes mellitus in rats. Diabetic rats with <85% of their basic levels in mechanical withdrawal threshold and thermal withdrawal latency were classified as DNP rats on day 14 after STZ injection. DNP rats were treated with ROS scavenger N-tert-Butyl-α-phenylnitrone (PBN, 100 mg·kg -1 ·d -1 ) or TXNIP small interfering ribonucleic acid (10 μg/d) once daily for 14 days. The level of ROS, protein levels of NLRP3, TXNIP, cysteinyl aspartate-specific proteinase-1 (caspase-1), interleukin-1β (IL-1β), NR2B phosphorylation at Tyr1472 (p-NR2B), total NR2B (t-NR2B), and distribution of NLRP3 in the spinal cord were examined. In vitro experiments, BV2 cells and PC12 cells were individually cultured and cocultured in a high-glucose environment (35 mmol/L D-glucose). The level of ROS and protein levels of NLRP3, TXNIP, caspase-1, and IL-1β in BV2 cells, and p-NR2B, t-NR2B in PC12 cells were detected. The level of ROS was detected by the flow cytometry approach. The protein levels were detected by the Western blot technique. The location of NLRP3 was observed by immunofluorescent staining. The interaction between TXNIP and NLRP3 was detected by coimmunoprecipitation assay. The level of spinal ROS increased in DNP rats. The mechanical allodynia and thermal hyperalgesia of DNP rats were alleviated after systemic administration of PBN. This administration decreased protein levels of NLRP3, TXNIP, caspase-1, IL-1β, and p-NR2B and the coupling of TXNIP to NLRP3 in spinal cords of DNP rats. Furthermore, knockdown of spinal TXNIP alleviated nociceptive hypersensitivity and decreased protein levels of NLRP3, TXNIP, caspase-1, IL-1β, and p-NR2B in DNP rats. The level of ROS and protein levels of NLRP3, TXNIP, caspase-1, IL-1β, the coupling of TXNIP to NLRP3, and the IL-1β secretion increased in BV2 cells, and the protein expression of p-NR2B increased in cocultured PC12 cells in a high-glucose environment. All of these in vitro effects were significantly blocked after treatment of PBN. Our findings suggest that spinal ROS can contribute to type 2 DNP through TXNIP-NLRP3-NR2B pathway.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-NMDAR2B Antibody, phosphoTyr 1472, Chemicon®, from rabbit