Merck
  • Apolipoprotein (a)/Lipoprotein(a)-Induced Oxidative-Inflammatory α7-nAChR/p38 MAPK/IL-6/RhoA-GTP Signaling Axis and M1 Macrophage Polarization Modulate Inflammation-Associated Development of Coronary Artery Spasm.
Apolipoprotein (a)/Lipoprotein(a)-Induced Oxidative-Inflammatory α7-nAChR/p38 MAPK/IL-6/RhoA-GTP Signaling Axis and M1 Macrophage Polarization Modulate Inflammation-Associated Development of Coronary Artery Spasm.
Oxidative medicine and cellular longevity (2022-02-01)
Yen-Kuang Lin, Chi-Tai Yeh, Kuang-Tai Kuo, Iat-Hang Fong, Vijesh Kumar Yadav, Nicholas G Kounis, Patrick Hu, Ming-Yow Hung
ABSTRACT
Apolipoprotein (a)/lipoprotein(a) (Lp(a)), a major carrier of oxidized phospholipids, and α7-nicotinic acetylcholine receptor (α7-nAChR) may play an important role in the development of coronary artery spasm (CAS). In CAS, the association between Lp(a) and the α7-nAChR-modulated inflammatory macrophage polarization and activation and smooth muscle cell dysfunction remains unknown. We investigated the relevance of Lp(a)/α7-nAChR signaling in patient monocyte-derived macrophages and human coronary artery smooth muscle cells (HCASMCs) using expression profile correlation analyses, fluorescence-assisted cell sorting flow cytometry, immunoblotting, quantitative real-time polymerase chain reaction, and clinicopathological analyses. There are increased serum Lp(a) levels (3.98-fold, p = 0.011) and macrophage population (3.30-fold, p = 0.013) in patients with CAS compared with patients without CAS. Serum Lp(a) level was positively correlated with high-sensitivity C-reactive protein (r 2 = 0.48, p < 0.01), IL-6 (r 2 = 0.38, p = 0.03), and α7-nAChR (r 2 = 0.45, p < 0.01) in patients with CAS, but not in patients without CAS. Compared with untreated or low-density lipoprotein- (LDL-) treated macrophages, Lp(a)-treated macrophages exhibited markedly enhanced α7-nAChR mRNA expression (p < 0.01) and activity (p < 0.01), in vitro and ex vivo. Lp(a) but not LDL preferentially induced CD80+ macrophage (M1) polarization and reduced the inducible nitric oxide synthase expression and the subsequent NO production. While shRNA-mediated loss of α7-nAChR function reduced the Lp(a)-induced CD80+ macrophage pool, both shRNA and anti-IL-6 receptor tocilizumab suppressed Lp(a)-upregulated α7-nAChR, p-p38 MAPK, IL-6, and RhoA-GTP protein expression levels in cultures of patient monocyte-derived macrophages and HCASMCs. Elevated Lp(a) levels upregulate α7-nAChR/IL-6/p38 MAPK signaling in macrophages of CAS patients and HCASMC, suggesting that Lp(a)-triggered inflammation mediates CAS through α7-nAChR/p38 MAPK/IL-6/RhoA-GTP signaling induction, macrophage M1 polarization, and HCASMC activation.
Sigma-Aldrich
Interferon-γ human, IFN-gamma, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-6, Human, Recombinant, E. coli
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
C Reactive Protein human, recombinant, expressed in E. coli, liquid
Sigma-Aldrich
Interleukin-4 human, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture