Merck
  • A detrimental role of NLRP6 in host iron metabolism during Salmonella infection.
A detrimental role of NLRP6 in host iron metabolism during Salmonella infection.
Redox biology (2021-12-24)
Qifeng Deng, Sidi Yang, Lanqing Sun, Kai Huang, Kedi Dong, Yuan Zhu, Yu Cao, Yuanyuan Li, Shuyan Wu, Rui Huang
ABSTRACT
Maintaining host iron homeostasis is an essential component of nutritional immunity responsible for sequestrating iron from pathogens and controlling infection. Nucleotide-oligomerization domain-like receptors (NLRs) contribute to cytoplasmic sensing and antimicrobial response orchestration. However, it remains unknown whether and how NLRs may regulate host iron metabolism, an important component of nutritional immunity. Here, we demonstrated that NLRP6, a member of the NLR family, has an unconventional role in regulating host iron metabolism that perturbs host resistance to bacterial infection. NLRP6 deficiency is advantageous for maintaining cellular iron homeostasis in both macrophages and enterocytes through increasing the unique iron exporter ferroportin-mediated iron efflux in a nuclear factor erythroid-derived 2-related factor 2 (NRF2)-dependent manner. Additional studies uncovered a novel mechanism underlying NRF2 regulation and operating through NLRP6/AKT interaction and that causes a decrease in AKT phosphorylation, which in turn reduces NRF2 nuclear translocation. In the absence of NLRP6, increased AKT activation promotes NRF2/KEAP1 dissociation via increasing mTOR-mediated p62 phosphorylation and downregulates KEAP1 transcription by promoting FOXO3A phosphorylation. Together, our observations provide new insights into the mechanism of nutritional immunity by revealing a novel function of NLRP6 in regulating iron metabolism, and suggest NLRP6 as a therapeutic target for limiting bacterial iron acquisition.
Sigma-Aldrich
MONOCLONAL ANTI-BETA-ACTIN antibody produced in mouse, clone 137CT26.1.1, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
ANTI-MOUSE NLRP6 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-GFP antibody produced in rabbit, affinity isolated antibody