Merck
  • Bionanoconjugation via click chemistry: The creation of functional hybrids of lipases and gold nanoparticles.
Bionanoconjugation via click chemistry: The creation of functional hybrids of lipases and gold nanoparticles.
Bioconjugate chemistry (2006-11-16)
Jennifer L Brennan, Nikos S Hatzakis, T Robert Tshikhudo, Nijole Dirvianskyte, Valdemaras Razumas, Shamkant Patkar, Jesper Vind, Allan Svendsen, Roeland J M Nolte, Alan E Rowan, Mathias Brust
ABSTRACT
A simple and versatile method for the preparation of functional enzyme-gold nanoparticle conjugates using "click" chemistry has been developed. In a copper-catalyzed 1,2,3-triazole cycloaddition, an acetylene-functionalized Thermomyces lanuginosus lipase has been attached to azide-functionalized water-soluble gold nanoparticles under retention of enzymatic activity. The products have been characterized by gel electrophoresis and a fluorometric lipase activity assay. It is estimated that the equivalent of approximately seven fully active lipase molecules are attached to each nanoparticle.
Sigma-Aldrich
Gold nanoparticles, 40 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 30 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 50 nm diameter, azide functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 60 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 20 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 5 nm diameter, azide functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 15 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 90 nm diameter, azide functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 80 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 70 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 100 nm diameter, azide functionalized, 1ml, OD 50, in water