Merck
  • Nitric oxide-coupled signaling in odor elicited molecular events in the olfactory center of the terrestrial snail, Helix pomatia.
Nitric oxide-coupled signaling in odor elicited molecular events in the olfactory center of the terrestrial snail, Helix pomatia.
Cellular signalling (2016-11-26)
Zoltán Serfőző, Kálmán Nacsa, Zoltán Veréb, Izabella Battonyai, Csaba Hegedűs, Csilla Balogh, Károly Elekes
ABSTRACT
Olfaction, a chemosensory modality, plays a pivotal role in the orientation and behavior of invertebrates. The central olfactory processing unit in terrestrial stylomatophoran snails is the procerebrum, which contains NO synthesizing interneurons, whose oscillatory currents are believed to be the base of odor evoked memory formation. Nevertheless, in this model the up- and downstream events of molecular cascades that trigger and follow NO release, respectively, have not been studied. Immunocytochemistry and flow cytometry studies performed on procerebral neural perikarya isolated from the snail Helix pomatia revealed cell populations with discrete DAF-2 fluorescence, indicating the release of different amounts of NO. Glutamate increased the intensity of DAF-2 fluorescence, and the number of DAF-2 positive non-bursting interneurons, through a mechanism likely to involve an NMDA-like receptor. Similarly to glutamate, NO activation induced an increase in intracellular cGMP levels through activation of soluble guanylyl cyclase. Immunohistochemical localization of proteins possessing the phosphorylated target sequence of AGC family kinases (RXXS/T-P), among them protein kinase A (RRXS/T-P), showed striking similarities to the distribution of NOS/cGMP. Activators of cyclic nucleotide synthesis increased the AGC-kinase-dependent phosphorylation of discrete proteins with 28, 45, and 55kDamw. Importantly, exposure of snails to an attractive odorant induced hyperphosphorylation of the 28kDa protein, and increased levels of cGMP synthesis. Protein S-nitrosylation and intercellular activation of protein kinase G were also suggested as alternative components of NO signaling in the snail procerebrum. The present results from Helix pomatia indicate an important role for procerebrum NO/cGMP/PKA signaling pathways in the regulation of olfactory (food-finding) behavior.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt, vial of 0.30-0.36 mg
Sigma-Aldrich
S-Nitrosoglutathione, ≥97%
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
4,5-Diaminofluorescein diacetate solution, 5 mM in DMSO, ≥97% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder