Merck
  • Non-canonical Targets of HIF1a Impair Oligodendrocyte Progenitor Cell Function.
Non-canonical Targets of HIF1a Impair Oligodendrocyte Progenitor Cell Function.
Cell stem cell (2020-10-23)
Kevin C Allan, Lucille R Hu, Marissa A Scavuzzo, Andrew R Morton, Artur S Gevorgyan, Erin F Cohn, Benjamin L L Clayton, Ilya R Bederman, Stevephen Hung, Cynthia F Bartels, Mayur Madhavan, Paul J Tesar
ABSTRACT
Mammalian cells respond to insufficient oxygen through transcriptional regulators called hypoxia-inducible factors (HIFs). Although transiently protective, prolonged HIF activity drives distinct pathological responses in different tissues. Using a model of chronic HIF1a accumulation in pluripotent-stem-cell-derived oligodendrocyte progenitors (OPCs), we demonstrate that HIF1a activates non-canonical targets to impair generation of oligodendrocytes from OPCs. HIF1a activated a unique set of genes in OPCs through interaction with the OPC-specific transcription factor OLIG2. Non-canonical targets, including Ascl2 and Dlx3, were sufficient to block differentiation through suppression of the oligodendrocyte regulator Sox10. Chemical screening revealed that inhibition of MEK/ERK signaling overcame the HIF1a-mediated block in oligodendrocyte generation by restoring Sox10 expression without affecting canonical HIF1a activity. MEK/ERK inhibition also drove oligodendrocyte formation in hypoxic regions of human oligocortical spheroids. This work defines mechanisms by which HIF1a impairs oligodendrocyte formation and establishes that cell-type-specific HIF1a targets perturb cell function in response to low oxygen.
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
Anti-Achaete Scute homolog 2 Antibody, clone 8F1, clone 8F1, from mouse
Sigma-Aldrich
Anti-A2B5 Antibody, clone A2B5-105, clone A2B5-105, Chemicon®, from mouse
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
LDN193189 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-NG2, Alexa Fluor488 Conjugate Antibody, from rabbit, ALEXA FLUOR 488
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse