Merck
Distinct lipid metabolic dysregulation in asymptomatic COVID-19.
iScience (2021-08-17)
Yanhong Hao, Zheng Zhang, Guifang Feng, Moran Chen, Qiongqiong Wan, Jie Lin, Liang Wu, Wenjing Nie, Suming Chen
ABSTRACT
Asymptomatic infection is a big challenge in curbing the spread of COVID-19. However, its identification and pathogenesis elucidation remain issues. Here, by performing comprehensive lipidomic characterization of serum samples from 89 asymptomatic COVID-19 patients and 178 healthy controls, we screened out a panel of 15 key lipids that could accurately identify asymptomatic patients using a new ensemble learning model based on stacking strategy with a voting algorithm. This strategy provided a high accuracy of 96.0% with only 3.6% false positive rate and 4.8% false negative rate. More importantly, the unique lipid metabolic dysregulation was revealed, especially the enhanced synthesis of membrane phospholipids, altered sphingolipids homeostasis, and differential fatty acids metabolic pattern, implicating the specific host immune, inflammatory, and antiviral responses in asymptomatic COVID-19. This study provides a potential prediagnostic method for asymptomatic COVID-19 and molecular clues for the pathogenesis and therapy of this disease.
Sigma-Aldrich
Myristic acid, ≥98.0% (GC)
Sigma-Aldrich
Behenic acid, 99%
Supelco
all-cis-4,7,10,13,16-Docosapentaenoic acid, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)