Merck
  • High throughput screening data for a case study of CHO cell culture process development.
High throughput screening data for a case study of CHO cell culture process development.
Data in brief (2021-10-30)
Qin He, Jianlin Xu, Erik Vandermark, Jun Tian, Yueming Qian, Cynthia Chelius, Jongchan Lee, Michael C Borys, Zheng Jian Li, Girish Pendse
ABSTRACT
In this article, we present four sets of data from high-throughput screening (HTS) studies of different chemically defined media using an industrially relevant Chinese hamster ovary (CHO) cell line. While complex hydrolysate media was used in the early phase process development and manufacturing of a monoclonal antibody (mAb), here we seek to determine an appropriate chemically defined media for late phase process development. Over 150 combinations of chemically defined basal media, feed media, and basal and feed media supplements, such as polyphenolic flavonoid antioxidants (including rosmarinic acid (RA)), were evaluated in four HTS studies to replace the complex hydrolysate media. Specifically, these four screening studies incorporated custom design of experiment (DOE), one-factor-at-a-time (OFAT), and definitive screening design methodologies for titer improvement. Titer was improved two fold compared to the early phase process using the addition of RA to chemically defined media. This dataset exemplifies how HTS can be used as an effective approach to systematically and statistically determine media composition and supplementation to increase mAb titer. These data were presented in connection with a published paper [1].
Product Description
USP
(+)-Catechin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)