Merck
  • Microfluidic tumor-on-a-chip model to evaluate the role of tumor environmental stress on NK cell exhaustion.
Microfluidic tumor-on-a-chip model to evaluate the role of tumor environmental stress on NK cell exhaustion.
Science advances (2021-02-19)
Jose M Ayuso, Shujah Rehman, Maria Virumbrales-Munoz, Patrick H McMinn, Peter Geiger, Cate Fitzgerald, Tiffany Heaster, Melissa C Skala, David J Beebe
ABSTRACT
Solid tumors generate a suppressive environment that imposes an overwhelming burden on the immune system. Nutrient depletion, waste product accumulation, hypoxia, and pH acidification severely compromise the capacity of effector immune cells such as T and natural killer (NK) cells to destroy cancer cells. However, the specific molecular mechanisms driving immune suppression, as well as the capacity of immune cells to adapt to the suppressive environment, are not completely understood. Thus, here, we used an in vitro microfluidic tumor-on-a-chip platform to evaluate how NK cells respond to the tumor-induced suppressive environment. The results demonstrated that the suppressive environment created by the tumor gradually eroded NK cell cytotoxic capacity, leading to compromised NK cell surveillance and tumor tolerance. Further, NK cell exhaustion persisted for an extended period of time after removing NK cells from the microfluidic platform. Last, the addition of checkpoint inhibitors and immunomodulatory agents alleviated NK cell exhaustion.
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline