Merck
  • An adhesion G protein-coupled receptor is required in cartilaginous and dense connective tissues to maintain spine alignment.
An adhesion G protein-coupled receptor is required in cartilaginous and dense connective tissues to maintain spine alignment.
eLife (2021-07-29)
Zhaoyang Liu, Amro A Hussien, Yunjia Wang, Terry Heckmann, Roberto Gonzalez, Courtney M Karner, Jess G Snedeker, Ryan S Gray
ABSTRACT
Adolescent idiopathic scoliosis (AIS) is the most common spine disorder affecting children worldwide, yet little is known about the pathogenesis of this disorder. Here, we demonstrate that genetic regulation of structural components of the axial skeleton, the intervertebral discs, and dense connective tissues (i.e., ligaments and tendons) is essential for the maintenance of spinal alignment. We show that the adhesion G protein-coupled receptor ADGRG6, previously implicated in human AIS association studies, is required in these tissues to maintain typical spine alignment in mice. Furthermore, we show that ADGRG6 regulates biomechanical properties of tendon and stimulates CREB signaling governing gene expression in cartilaginous tissues of the spine. Treatment with a cAMP agonist could mirror aspects of receptor function in culture, thus defining core pathways for regulating these axial cartilaginous and connective tissues. As ADGRG6 is a key gene involved in human AIS, these findings open up novel therapeutic opportunities for human scoliosis.
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
TGF-β3 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE)
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Tamoxifen, ≥99%