Merck
  • Collagen I triggers directional migration, invasion and matrix remodeling of stroma cells in a 3D spheroid model of endometriosis.
Collagen I triggers directional migration, invasion and matrix remodeling of stroma cells in a 3D spheroid model of endometriosis.
Scientific reports (2021-02-20)
Anna Stejskalová, Victoria Fincke, Melissa Nowak, Yvonne Schmidt, Katrin Borrmann, Marie-Kristin von Wahlde, Sebastian D Schäfer, Ludwig Kiesel, Burkhard Greve, Martin Götte
ABSTRACT
Endometriosis is a painful gynecological condition characterized by ectopic growth of endometrial cells. Little is known about its pathogenesis, which is partially due to a lack of suitable experimental models. Here, we use endometrial stromal (St-T1b), primary endometriotic stromal, epithelial endometriotic (12Z) and co-culture (1:1 St-T1b:12Z) spheroids to mimic the architecture of endometrium, and either collagen I or Matrigel to model ectopic locations. Stromal spheroids, but not single cells, assumed coordinated directional migration followed by matrix remodeling of collagen I on day 5 or 7, resembling ectopic lesions. While generally a higher area fold increase of spheroids occurred on collagen I compared to Matrigel, directional migration was not observed in co-culture or in 12Z cells. The fold increase in area on collagen I was significantly reduced by MMP inhibition in stromal but not 12Z cells. Inhibiting ROCK signalling responsible for actomyosin contraction increased the fold increase of area and metabolic activity compared to untreated controls on Matrigel. The number of protrusions emanating from 12Z spheroids on Matrigel was decreased by microRNA miR-200b and increased by miR-145. This study demonstrates that spheroid assay is a promising pre-clinical tool that can be used to evaluate small molecule drugs and microRNA-based therapeutics for endometriosis.
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, 10×, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
StableCell DMEM - high glucose, With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
FBS Superior, Supplemented FBS, EU approved
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
MCDB 105 Medium (500 ml)
Sigma-Aldrich
NNGH, ≥98% (HPLC)
Sigma-Aldrich
DAPI, dilactate, ≥98% (HPLC)