Merck
  • Reduction Mechanism of Transition Metal Oxide Particles in Thermally Induced Nanobubbles during Pulsed Laser Melting in Ethanol.
Reduction Mechanism of Transition Metal Oxide Particles in Thermally Induced Nanobubbles during Pulsed Laser Melting in Ethanol.
Chemphyschem : a European journal of chemical physics and physical chemistry (2021-01-27)
Kentaro Suehara, Ryosuke Takai, Yoshie Ishikawa, Naoto Koshizaki, Kazunobu Omura, Harunori Nagata, Yuji Yamauchi
ABSTRACT
Pulsed laser melting in liquid (PLML) is a technique to fabricate spherical submicrometer particles (SMPs) wherein nanosecond pulsed laser (several tens to several hundreds of mJ pulse-1 cm-2 ) irradiates raw particles dispersed in liquid. Raw particles are transiently heated above the melting point to form spherical particles, which enables pulsed heating of surrounding liquid to form thermally induced bubbles by liquid vaporization. These transient bubbles play an important role as a thermal barrier to rapidly heat the particle. Reduced SMPs are generated from raw metal-oxide nanoparticles by PLML process in ethanol. This reduction cannot be explained by high-temperature thermal decomposition, but by mediation of molecules decomposed from ethanol. Computational simulations of ethanol decomposition by pulsed heating for 100 ns at the temperature 1000-4000 K revealed that ethylene is generated as the main product. Gibbs free energies of oxide reduction reactions mediated by ethylene greatly decreased compared to those without ethylene mediation. This explanation can be applied to reductive SMP formation from various transition metal oxides by PLML.
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Cobalt(II,III) oxide, nanopowder, <50 nm particle size (SEM), 99.5% trace metals basis
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Nickel(II) oxide, nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis