Merck
  • Kv1.3 voltage-gated potassium channels link cellular respiration to proliferation through a non-conducting mechanism.
Kv1.3 voltage-gated potassium channels link cellular respiration to proliferation through a non-conducting mechanism.
Cell death & disease (2021-04-09)
Faye L Styles, Moza M Al-Owais, Jason L Scragg, Eulashini Chuntharpursat-Bon, Nishani T Hettiarachchi, Jonathan D Lippiat, Aisling Minard, Robin S Bon, Karen Porter, Piruthivi Sukumar, Chris Peers, Lee D Roberts
ABSTRACT
Cellular energy metabolism is fundamental for all biological functions. Cellular proliferation requires extensive metabolic reprogramming and has a high energy demand. The Kv1.3 voltage-gated potassium channel drives cellular proliferation. Kv1.3 channels localise to mitochondria. Using high-resolution respirometry, we show Kv1.3 channels increase oxidative phosphorylation, independently of redox balance, mitochondrial membrane potential or calcium signalling. Kv1.3-induced respiration increased reactive oxygen species production. Reducing reactive oxygen concentrations inhibited Kv1.3-induced proliferation. Selective Kv1.3 mutation identified that channel-induced respiration required an intact voltage sensor and C-terminal ERK1/2 phosphorylation site, but is channel pore independent. We show Kv1.3 channels regulate respiration through a non-conducting mechanism to generate reactive oxygen species which drive proliferation. This study identifies a Kv1.3-mediated mechanism underlying the metabolic regulation of proliferation, which may provide a therapeutic target for diseases characterised by dysfunctional proliferation and cell growth.
Sigma-Aldrich
PAPTP trifluoroacetate, ≥98% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture