Merck
BRAF inhibition protects against hearing loss in mice.
Science advances (2020-12-04)
Matthew A Ingersoll, Emma A Malloy, Lauryn E Caster, Eva M Holland, Zhenhang Xu, Marisa Zallocchi, Duane Currier, Huizhan Liu, David Z Z He, Jaeki Min, Taosheng Chen, Jian Zuo, Tal Teitz
ABSTRACT
Hearing loss caused by noise, aging, antibiotics, and chemotherapy affects 10% of the world population, yet there are no Food and Drug Administration (FDA)-approved drugs to prevent it. Here, we screened 162 small-molecule kinase-specific inhibitors for reduction of cisplatin toxicity in an inner ear cell line and identified dabrafenib (TAFINLAR), a BRAF kinase inhibitor FDA-approved for cancer treatment. Dabrafenib and six additional kinase inhibitors in the BRAF/MEK/ERK cellular pathway mitigated cisplatin-induced hair cell death in the cell line and mouse cochlear explants. In adult mice, oral delivery of dabrafenib repressed ERK phosphorylation in cochlear cells, and protected from cisplatin- and noise-induced hearing loss. Full protection was achieved in mice with co-treatment with oral AZD5438, a CDK2 kinase inhibitor. Our study explores a previously unidentified cellular pathway and molecular target BRAF kinase for otoprotection and may advance dabrafenib into clinics to benefit patients with cisplatin- and noise-induced ototoxicity.
Sigma-Aldrich
TWEEN® 80, viscous liquid
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Sigma-Aldrich
Ampicillin sodium salt, Ready Made Solution, 100 mg/mL, 0.2 μm filtered
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%
Sigma-Aldrich
Kollisolv® PEG E 300