Merck
  • Backbone cyclic peptidomimetic melanocortin-4 receptor agonist as a novel orally administrated drug lead for treating obesity.
Backbone cyclic peptidomimetic melanocortin-4 receptor agonist as a novel orally administrated drug lead for treating obesity.
Journal of medicinal chemistry (2008-01-29)
Shmuel Hess, Yaniv Linde, Oded Ovadia, Eli Safrai, Deborah E Shalev, Avi Swed, Efrat Halbfinger, Tair Lapidot, Ilan Winkler, Yael Gabinet, Avi Faier, Dana Yarden, Zhimin Xiang, Federico P Portillo, Carrie Haskell-Luevano, Chaim Gilon, Amnon Hoffman
ABSTRACT
The tetrapeptide sequence His-Phe-Arg-Trp, derived from melanocyte-stimulating hormone (alphaMSH) and its analogs, causes a decrease in food intake and elevates energy utilization upon binding to the melanocortin-4 receptor (MC4R). To utilize this sequence as an effective agent for treating obesity, we improved its metabolic stability and intestinal permeability by synthesizing a library of backbone cyclic peptidomimetic derivatives. One analog, peptide 1 (BL3020-1), was selected according to its selectivity in activating the MC4R, its favorable transcellular penetration through enterocytes and its enhanced intestinal metabolic stability. This peptide was detected in the brain following oral administration to rats. A single oral dose of 0.5 mg/kg in mice led to reduced food consumption (up to 48% vs the control group) that lasted for 5 h. Repetitive once daily oral dosing (0.5 mg/kg/day) for 12 days reduced weight gain. Backbone cyclization was shown to produce a potential drug lead for treating obesity.
Product Description
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)