Merck
  • Contamination, Decomposition, and Formation of N-Nitrosodimethylamine in Water Samples at the ng/L Level of Determination.
Contamination, Decomposition, and Formation of N-Nitrosodimethylamine in Water Samples at the ng/L Level of Determination.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2020-07-28)
Hitoshi Kodamatani, Kenta Sugihara, Taketo Tanisue, Ryo Kanzaki, Takashi Tomiyasu
ABSTRACT
An ultra-sensitive analytical system that can determine the concentration of N-nitrosamines at the ng/L level without preconcentration was used to investigate the contamination, decomposition, and formation of N-nitrosodimethylamine (NDMA) and other N-nitrosamines in water samples during general analytical procedures. A preliminary experiment was performed to estimate the NDMA concentrations in ambient air. Since the air samples contained NDMA at concentrations in the range of 2.0 - 10.7 ng/m3, ambient air was identified as the source of NDMA contamination in water samples. We directly confirmed that the concentration of aqueous 10-ng/L NDMA samples stored in clear glass bottles decreased upon exposure to sunlight. Thus, to maintain the N-nitrosamine concentration, such samples must always be protected from sunlight during sampling. The existence of N-nitrosamines in experimental reagents, such as ranitidine and sodium hypochlorite solutions, was also confirmed, as was the formation of NDMA on an activated carbon solid-phase extraction cartridge.
Product Description
Molsidomine impurity B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-Ethylmethylamine, 97%