Merck
PCNA Unloading Is Negatively Regulated by BET Proteins.
Cell reports (2019-12-26)
Mi-Sun Kang, Jinwoo Kim, Eunjin Ryu, Na Young Ha, Sunyoung Hwang, Byung-Gyu Kim, Jae Sun Ra, Yeong Jae Kim, Jung Me Hwang, Kyungjae Myung, Sukhyun Kang
ABSTRACT
Proliferating cell nuclear antigen (PCNA) is a DNA clamp essential for DNA replication. During DNA synthesis, PCNA is continuously loaded onto and unloaded from DNA. PCNA recruits various proteins to nascent DNA to facilitate chromosome duplication. Therefore, timely PCNA unloading is crucial for high-fidelity DNA replication. The ATAD5-RFC-like complex (ATAD5-RLC) unloads PCNA from replicated DNA. It is unclear how ATAD5-RLC activity is regulated to prevent premature PCNA unloading. Here, we find that BRD4, an acetyl-histone-binding chromatin reader, inhibits the PCNA-unloading activity of ATAD5-RLC. The BRD4 ET domain interacts with a region upstream of the ATAD5 PCNA-unloading domain. BRD4-ATAD5 binds to acetyl-histones in nascent chromatin. BRD4 release from chromatin correlates with PCNA unloading. Disruption of the interaction between BRD4 and acetyl-histones or between BRD4 and ATAD5 reduces the PCNA amount on chromatin. In contrast, the overexpression of BRD4 increases the amount of chromatin-bound PCNA. Thus, acetyl-histone-bound BRD4 fine-tunes PCNA unloading from nascent DNA.
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
d-Desthiobiotin, ≥98% (TLC)
Millipore
Anti-V5 Agarose Affinity Gel antibody produced in mouse, purified immunoglobulin, clone V5-10
Sigma-Aldrich
Acetyl-Histone H4 Antibody Set, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, serum, Upstate®
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Monoclonal Anti-S Tag antibody produced in mouse, clone GT727, affinity isolated antibody
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
(+/-)-JQ1, ≥98% (HPLC)
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Anti-V5 antibody produced in rabbit, IgG fraction of antiserum
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution