Merck
  • Macrophages Maintain Epithelium Integrity by Limiting Fungal Product Absorption.
Macrophages Maintain Epithelium Integrity by Limiting Fungal Product Absorption.
Cell (2020-09-25)
Aleksandra S Chikina, Francesca Nadalin, Mathieu Maurin, Mabel San-Roman, Thibault Thomas-Bonafos, Xin V Li, Sonia Lameiras, Sylvain Baulande, Sandrine Henri, Bernard Malissen, Livia Lacerda Mariano, Jorge Barbazan, J Magarian Blander, Iliyan D Iliev, Danijela Matic Vignjevic, Ana-Maria Lennon-Duménil
ABSTRACT
The colon is primarily responsible for absorbing fluids. It contains a large number of microorganisms including fungi, which are enriched in its distal segment. The colonic mucosa must therefore tightly regulate fluid influx to control absorption of fungal metabolites, which can be toxic to epithelial cells and lead to barrier dysfunction. How this is achieved remains unknown. Here, we describe a mechanism by which the innate immune system allows rapid quality check of absorbed fluids to avoid intoxication of colonocytes. This mechanism relies on a population of distal colon macrophages that are equipped with "balloon-like" protrusions (BLPs) inserted in the epithelium, which sample absorbed fluids. In the absence of macrophages or BLPs, epithelial cells keep absorbing fluids containing fungal products, leading to their death and subsequent loss of epithelial barrier integrity. These results reveal an unexpected and essential role of macrophages in the maintenance of colon-microbiota interactions in homeostasis. VIDEO ABSTRACT.
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
Sunflower seed oil from Helianthus annuus
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Supelco
Bisacodyl, analytical standard, for drug analysis
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Gliotoxin from Gliocladium fimbriatum
Sigma-Aldrich
Hanks′ Balanced Salt solution, 10 ×, Modified, without calcium, magnesium or sodium bicarbonate
Sigma-Aldrich
Gentamicin sulfate, meets EP, USP testing specifications
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)