Merck
  • Epitaxially Grown Films of Standing and Lying Pentacene Molecules on Cu(110) Surfaces.
Epitaxially Grown Films of Standing and Lying Pentacene Molecules on Cu(110) Surfaces.
Crystal growth & design (2011-04-12)
Tatjana Djuric, Thomas Ules, Heinz-Georg Flesch, Harald Plank, Quan Shen, Christian Teichert, Roland Resel, Michael G Ramsey
ABSTRACT
This study investigates epitaxially oriented pentacene films grown on Cu(110) surfaces crystallizing either in the "thin film" phase with standing molecules or in the "single crystal" structure with molecules lying with their long axes parallel to the substrate.
Sigma-Aldrich
Pentacene, triple-sublimed grade, ≥99.995% trace metals basis
Sigma-Aldrich
Pentacene, 99%
Sigma-Aldrich
Pentacene, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Pentacene, for fluorescence, >95.0%