Merck
  • New Platinum(II) Triazole Thiosemicarbazone Complexes: Analysis of Their Reactivity and Potential Antitumoral Action.
New Platinum(II) Triazole Thiosemicarbazone Complexes: Analysis of Their Reactivity and Potential Antitumoral Action.
Chembiochem : a European journal of chemical biology (2019-11-21)
Ana I Matesanz, Jorge M Herrero, Eva J Faraco, Leticia Cubo, Adoracion G Quiroga
ABSTRACT
The synthesis and characterization of three new platinum complexes, with 3,5-diacetyl-1,2,4-triazole bis(4-N-isopropylthiosemicarbazone) as a ligand, are reported. The specific conditions under which solvent coordination takes place are reported and the X-ray structure of the complex with one solvent molecule of dimethyl sulfoxide is resolved. Analysis of the reactivity of these platinum compounds aids in finding the best solution profile for biological investigations. Then, the interactions of the complexes with biological models, such as calf-thymus DNA, are studied by using UV spectroscopy and tracking the changes in electrophoretic mobility produced in the supercoiled plasmid DNA model. Initial screening of these potential antitumoral compounds indicates possible selective antitumoral action.
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture
Sigma-Aldrich
Insulin from bovine pancreas, γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder
Sigma-Aldrich
1 kb DNA Ladder, for DNA electrophoresis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)