Merck
  • CSF1R Is Required for Differentiation and Migration of Langerhans Cells and Langerhans Cell Histiocytosis.
CSF1R Is Required for Differentiation and Migration of Langerhans Cells and Langerhans Cell Histiocytosis.
Cancer immunology research (2020-04-03)
Silvia Lonardi, Sara Scutera, Sara Licini, Luisa Lorenzi, Anna Maria Cesinaro, Luisa Benerini Gatta, Carlotta Castagnoli, Daniele Bollero, Rosaria Sparti, Michela Tomaselli, Daniela Medicina, Federica Calzetti, Marco Antonio Cassatella, Fabio Facchetti, Tiziana Musso, William Vermi
ABSTRACT
Langerhans cell histiocytosis (LCH) is a rare disorder characterized by tissue accumulation of CD1a+CD207+ LCH cells. In LCH, somatic mutations of the BRAF V600E gene have been detected in tissue LCH cells, bone marrow CD34+ hematopoietic stem cells, circulating CD14+ monocytes, and BDCA1+ myeloid dendritic cells (DC). Targeting BRAF V600E in clonal Langerhans cells (LC) and their precursors is a potential treatment option for patients whose tumors have the mutation. The development of mouse macrophages and LCs is regulated by the CSF1 receptor (CSF1R). In patients with diffuse-type tenosynovial giant cell tumors, CSF1R inhibition depletes tumor-associated macrophages (TAM) with therapeutic efficacy; however, CSF1R signaling in LCs and LCH has not been investigated. We found through IHC and flow cytometry that CSF1R is normally expressed on human CD1a+CD207+ LCs in the epidermis and stratified epithelia. LCs that were differentiated from CD14+ monocytes, BDCA1+ DCs, and CD34+ cord blood progenitors expressed CSF1R that was downregulated upon maturation. Immature LCs migrated toward CSF1, but not IL34. Administration of the c-FMS/CSF1R kinase inhibitors GW2580 and BLZ945 significantly reduced human LC migration. In LCH clinical samples, LCH cells (including BRAF V600E cells) and TAMs retained high expression of CSF1R. We also detected the presence of transcripts for its ligand, CSF1, but not IL34, in all tested LCH cases. CSF1R and CSF1 expression in LCH, and their role in LC migration and differentiation, suggests CSF1R signaling blockade as a candidate rational approach for treatment of LCH, including the BRAF V600E and wild-type forms of the disease.
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
GW2580, ≥98% (HPLC)
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (HPLC), buffered aqueous solution
Roche
DNase I, grade II, from bovine pancreas