Merck
  • The omega-loop region of the human prothrombin gamma-carboxyglutamic acid domain penetrates anionic phospholipid membranes.
The omega-loop region of the human prothrombin gamma-carboxyglutamic acid domain penetrates anionic phospholipid membranes.
The Journal of biological chemistry (2001-04-20)
L A Falls, B C Furie, M Jacobs, B Furie, A C Rigby
ABSTRACT
The hydrophobic omega-loop within the prothrombin gamma-carboxyglutamic acid-rich (Gla) domain is important in membrane binding. The role of this region in membrane binding was investigated using a synthetic peptide, PT-(1-46)F4W, which includes the N-terminal 46 residues of human prothrombin with Phe-4 replaced by Trp providing a fluorescent probe. PT-(1-46)F4W and PT-(1-46) bind calcium ions and phospholipid membranes, and inhibit the prothrombinase complex. PT-(1-46)F4W, but not PT-(1-46), exhibits a blue shift (5 nm) and red-edge excitation shift (28 nm) in the presence of phosphatidylserine (PS)-containing vesicles, suggesting Trp-4 is located within the motionally restricted membrane interfacial region. PS-containing vesicles protect PT-(1-46)F4W, but not PT-(1-46), fluorescence from potassium iodide-induced quenching. Stern-Volmer analysis of the quenching of PT-(1-46)F4W in the presence and absence of 80% phosphatidylcholine/20% PS vesicles suggested that Trp-4 is positioned within the membrane and protected from aqueous quenching agents whereas Trp-41 remains solvent-accessible in the presence of PS-containing vesicles. Fluorescence quenching of membrane-bound PT-(1-46)F4W is optimal with 7- and 10-doxyl-labeled lipids, indicating that Trp-4 is inserted 5 to 7 A into the bilayer. This report demonstrates that the omega-loop region of prothrombin specifically interacts with PS-containing membranes within the interfacial membrane region.
Product Description
Avanti
16:0-10 Doxyl PC, Avanti Research - A Croda Brand 810603C
Avanti
16:0-10 Doxyl PC, Avanti Research - A Croda Brand 810603P, powder