Merck
  • CCDC57 Cooperates with Microtubules and Microcephaly Protein CEP63 and Regulates Centriole Duplication and Mitotic Progression.
CCDC57 Cooperates with Microtubules and Microcephaly Protein CEP63 and Regulates Centriole Duplication and Mitotic Progression.
Cell reports (2020-05-14)
H Kubra Gurkaslar, Efraim Culfa, Melis D Arslanhan, Mariana Lince-Faria, Elif Nur Firat-Karalar
ABSTRACT
Centrosomes function in key cellular processes ranging from cell division to cellular signaling. Their dysfunction is linked to cancer and developmental disorders. Here, we identify CCDC57 as a pleiotropic regulator of centriole duplication, mitosis, and ciliogenesis. Combining proximity mapping with superresolution imaging, we show that CCDC57 localizes to the proximal end of centrioles and interacts with the microcephaly protein CEP63, centriolar satellite proteins, and microtubules. Loss of CCDC57 causes defects in centriole duplication and results in a failure to localize CEP63 and CEP152 to the centrosome. Additionally, CCDC57 depletion perturbs mitotic progression both in wild-type and centriole-less cells. Importantly, its centrosome-targeting region is required for its interaction with CEP63 and functions during centriole duplication and cilium assembly, whereas the microtubule-targeting region is required for its mitotic functions. Together, our results identify CCDC57 as a critical interface between centrosome and microtubule-mediated cellular processes that are deregulated in microcephaly.
Sigma-Aldrich
MISSION® esiRNA, targeting human CCDC57
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human PCM1
Sigma-Aldrich
Anti-KIAA0753 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-CCDC57 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab4
Sigma-Aldrich
MISSION® esiRNA, targeting human CEP63