Merck
  • Propionate suppresses hepatic gluconeogenesis via GPR43/AMPK signaling pathway.
Propionate suppresses hepatic gluconeogenesis via GPR43/AMPK signaling pathway.
Archives of biochemistry and biophysics (2019-07-30)
Hiroki Yoshida, Megumi Ishii, Mitsugu Akagawa
ABSTRACT
Short-chain fatty acids (SCFAs) such as acetate, propionate, and butyrate are generated by gut microbial fermentation of dietary fiber. SCFAs may exert multiple beneficial effects on human lipid and glucose metabolism. However, their actions and underlying mechanisms are not fully elucidated. In this study, we examined the direct effects of propionate on hepatic glucose and lipid metabolism using human HepG2 hepatocytes. Here, we demonstrate that propionate at a physiologically-relevant concentration effectively suppresses palmitate-enhanced glucose production in HepG2 cells but does not affect intracellular neutral lipid levels. Our results indicated that propionate can decline in gluconeogenesis by down-regulation of glucose-6-phosphatase (G6Pase) and phosphoenolpyruvate carboxykinase (PEPCK) through activation of AMP-activated protein kinase (AMPK), which is a major regulator of the hepatic glucose metabolism. Mechanistic studies also revealed that propionate-stimulated AMPK phosphorylation can be ascribed to Ca2+/calmodulin-dependent protein kinase kinase β (CaMKKβ) activation in response to an increase in intracellular Ca2+ concentration. Moreover, siRNA-mediated knockdown of the propionate receptor GPR43 prevented propionate-inducible activation of AMPK and abrogates the gluconeogenesis-inhibitory action. Thus, our data indicate that the binding of propionate to hepatic GPR43 elicits CaMKKβ-dependent activation of AMPK through intracellular Ca2+ increase, leading to suppression of gluconeogenesis. The present study suggests the potential efficacy of propionate in preventive and therapeutic management of diabetes.
Sigma-Aldrich
MISSION® esiRNA, targeting human FFAR2
Sigma-Aldrich
Sodium palmitate, ≥98.5%