Merck
  • Direct Pharmacological Targeting of a Mitochondrial Ion Channel Selectively Kills Tumor Cells In Vivo.
Direct Pharmacological Targeting of a Mitochondrial Ion Channel Selectively Kills Tumor Cells In Vivo.
Cancer cell (2017-04-12)
Luigi Leanza, Matteo Romio, Katrin Anne Becker, Michele Azzolini, Livio Trentin, Antonella Managò, Elisa Venturini, Angela Zaccagnino, Andrea Mattarei, Luca Carraretto, Andrea Urbani, Stephanie Kadow, Lucia Biasutto, Veronica Martini, Filippo Severin, Roberta Peruzzo, Valentina Trimarco, Jan-Hendrik Egberts, Charlotte Hauser, Andrea Visentin, Gianpietro Semenzato, Holger Kalthoff, Mario Zoratti, Erich Gulbins, Cristina Paradisi, Ildiko Szabo
ABSTRACT
The potassium channel Kv1.3 is highly expressed in the mitochondria of various cancerous cells. Here we show that direct inhibition of Kv1.3 using two mitochondria-targeted inhibitors alters mitochondrial function and leads to reactive oxygen species (ROS)-mediated death of even chemoresistant cells independently of p53 status. These inhibitors killed 98% of ex vivo primary chronic B-lymphocytic leukemia tumor cells while sparing healthy B cells. In orthotopic mouse models of melanoma and pancreatic ductal adenocarcinoma, the compounds reduced tumor size by more than 90% and 60%, respectively, while sparing immune and cardiac functions. Our work provides direct evidence that specific pharmacological targeting of a mitochondrial potassium channel can lead to ROS-mediated selective apoptosis of cancer cells in vivo, without causing significant side effects.
Sigma-Aldrich
Staurosporine from Streptomyces sp., ≥98% (HPLC), film
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
PAPTP trifluoroacetate, ≥98% (HPLC)